Learn More
Aviva Systems Biology Corp RHBDD1 Antibody - C-terminal region (ARP60503_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60503_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RHBDD1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human RHBDD1. Alternate names: DKFZp547E052, MGC117258ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast
Catalog No. 50-287-065