Learn More
Aviva Systems Biology Corp RHBG Antibody - N-terminal region (ARP49502_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49502_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RHBG. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human RHBG. Alternate names: SLC42A2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-014-62958