Learn More
Aviva Systems Biology Corp RHCE Antibody - N-terminal region (ARP42567_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42567_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RHCE. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human RHCE. Alternate names: CD240CE, MGC103977, RH, RH30A, RHC, RHE, RHIXB, RHPI, Rh4, RhIVb(J), RhVI, RhVIIIApplicationWB. Species reactivity: Human
Catalog No. 50-014-58268