Learn More
Aviva Systems Biology Corp RHD Antibody - N-terminal region (ARP63086_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP63086_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RHD. It was validated on Western Blot. . Alternate names: CD240D, DIIIc, MGC165007, RH, RH30, RHCED, RHDVA(TT), RHDel, RHPII, RHXIII, Rh4, RhDCw, RhII, RhK562-II, RhPIApplicationWB. Species reactivity: Human
Catalog No. 50-287-869