Learn More
Medchemexpress LLC HY-124798 5mg Medchemexpress, Rheb inhibitor NR1 CAS:2216763-38-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1247985MG
Encompass_Preferred
Medchemexpress, HY-124798 5mg Rheb inhibitor NR1 CAS:2216763-38-9 Rheb inhibitor NR1 is a Rheb inhibitor with an IC50 of 2.1 µM in the Rheb-IVK assay. Rheb inhibitor NR1 also is a selective mTORC1 inhibitor. NR1 inhibits the phosphorylation of T389pS6K1 and increases the phosphorylation of S473pAKT in a dose-dependent manner. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9682