Learn More
Aviva Systems Biology Corp RHEBL1 Antibody - middle region (ARP52683_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52683_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RHEBL1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human RHEBL1. Alternate names: FLJ25797, MGC34869, RHEBL1cApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-60802