Learn More
Analytical Sales & Services RheBuild Kit for 2 Position, 6 Port High Pressure PEEK Switiching Valve (VXV7738-000)
Catalog No. 5000806040
Encompass
RheBuild Kit for 2 Position, 6 Port High Pressure PEEK Switiching Valve (VXV7738-000).\nIncludes: Stator Face Assembly, Rotor Seal, White Seal and Two Tools\n\nQty/Pkg: EACH
Catalog No. 50-008-06040 Supplier Analytical Sales & Services Supplier No. PD7738
RheBuild Kit for 2 Position, 6 Port High Pressure PEEK Switiching Valve (VXV7738-000).\nIncludes: Stator Face Assembly, Rotor Seal, White Seal and Two Tools\n\nQty/Pkg: EACH