Learn More
Analytical Sales & Services RheBuild Kit for 2 Position, 6 Port High Pressure PEEK Switiching Valve (VXV7788-000)
Catalog No. 5000805788
Encompass
RheBuild Kit for 2 Position, 6 Port High Pressure PEEK Switiching Valve (VXV7788-000)
Includes: Stator Face Assembly Rotor Seal White Seal and Two Tools
Qty/Pkg: EACH
Catalog No. 50-008-05788 Supplier Analytical Sales & Services Supplier No. 7788
RheBuild Kit for 2 Position, 6 Port High Pressure PEEK Switiching Valve (VXV7788-000)
Includes: Stator Face Assembly Rotor Seal White Seal and Two Tools
Qty/Pkg: EACH