Learn More
Analytical Sales & Services RheBuild Kit for 7010
Catalog No. 5000804771
Encompass
RheBuild Kit for 7010. Contains: Replacement Rotor Seals, Stator Face Assembly, Isolation Seal, Needle Guide, Needle Port Cleaner and Two Hex Nut Keys
Qty/Pkg: EACH
Catalog No. 50-008-04771 Supplier Analytical Sales & Services Supplier No. 470109974
RheBuild Kit for 7010. Contains: Replacement Rotor Seals, Stator Face Assembly, Isolation Seal, Needle Guide, Needle Port Cleaner and Two Hex Nut Keys
Qty/Pkg: EACH