Learn More
Analytical Sales & Services RheBuild Kit for 7125 7126
Catalog No. 5000805520
Encompass
RheBuild Kit for 7125, 7126. Contains: Replacement Rotor Seals, Stator Face Assembly, Isolation Seal, Needle Guide, Needle Port Cleaner and Two Hex Nut Keys
Qty/Pkg: EACH
Catalog No. 50-008-05520 Supplier Analytical Sales & Services Supplier No. 471259994
RheBuild Kit for 7125, 7126. Contains: Replacement Rotor Seals, Stator Face Assembly, Isolation Seal, Needle Guide, Needle Port Cleaner and Two Hex Nut Keys
Qty/Pkg: EACH