Learn More
Analytical Sales & Services RheBuild Kit for 7410
Catalog No. 5000804296
Encompass
RheBuild Kit for 7410. Contains: Replacement Rotor Seals, Stator Face Assembly, Isolation Seal, Needle Guide, Needle Port Cleaner and Two Hex Nut Keys
Qty/Pkg: EACH
Catalog No. 50-008-04296 Supplier Analytical Sales & Services Supplier No. 474109994
RheBuild Kit for 7410. Contains: Replacement Rotor Seals, Stator Face Assembly, Isolation Seal, Needle Guide, Needle Port Cleaner and Two Hex Nut Keys
Qty/Pkg: EACH