Learn More
Analytical Sales & Services RheBuild Kit for 7520, 7526
Catalog No. 5000804905
Encompass
RheBuild Kit for 7520, 7526. Contains: Replacement Rotor Seals, Stator Face Assembl, Isolation Seal, Needle Guide, Needle Port Cleaner and Two Hex Nut Keys
Qty/Pkg: EACH
Catalog No. 50-008-04905 Supplier Analytical Sales & Services Supplier No. 475209994
RheBuild Kit for 7520, 7526. Contains: Replacement Rotor Seals, Stator Face Assembl, Isolation Seal, Needle Guide, Needle Port Cleaner and Two Hex Nut Keys
Qty/Pkg: EACH