Learn More
Analytical Sales & Services RheBuild Kit for 7650, 9650
Catalog No. 5000804918
Encompass
RheBuild Kit for 7650, 9650\n\nContains: Replacement Rotor Seals, Stator Face Assembly, Isolation Seal, Needle Guide, Needle Port Cleaner and Two Hex Nut Keys
Catalog No. 50-008-04918 Supplier Analytical Sales & Services Supplier No. 496509994
RheBuild Kit for 7650, 9650\n\nContains: Replacement Rotor Seals, Stator Face Assembly, Isolation Seal, Needle Guide, Needle Port Cleaner and Two Hex Nut Keys