Learn More
Analytical Sales & Services RheBuild Kit for 7750 Series
Catalog No. 5000805639
Encompass
RheBuild Kit for 7750 Series. Contains: Replacement Rotor Seal, Stator Face Assembly, Isolation Seal, Needle Guide, Needle Port Cleaner and Two Hex Nut Keys
Qty/Pkg: EACH
Catalog No. 50-008-05639 Supplier Analytical Sales & Services Supplier No. 477509994
Add to Cart
Add to Cart
RheBuild Kit for 7750 Series. Contains: Replacement Rotor Seal, Stator Face Assembly, Isolation Seal, Needle Guide, Needle Port Cleaner and Two Hex Nut Keys
Qty/Pkg: EACH