Learn More
Analytical Sales & Services RheBuild Kit for 9125, 9126
Catalog No. 5000806937
Encompass
RheBuild Kit for 9125, 9126\n\nContains: Replacement Rotor Seals, Stator Face Assembly, Isolation Seal, Needle Guide, Needle Port Cleaner and Two Hex Nut Keys
Catalog No. 50-008-06937 Supplier Analytical Sales & Services Supplier No. 491259994
RheBuild Kit for 9125, 9126\n\nContains: Replacement Rotor Seals, Stator Face Assembly, Isolation Seal, Needle Guide, Needle Port Cleaner and Two Hex Nut Keys