Learn More
Analytical Sales & Services RheBuild Kit for 9725, 9725I
Catalog No. 5000806938
Encompass
RheBuild Kit for 9725, 9725I. Contains: Replacement Rotor Seals, Stator Face Assembly, Isolation Seal, Needle Guide, Needle Port Cleaner and Two Hex Nut Keys
Qty/Pkg: EACH
Catalog No. 50-008-06938 Supplier Analytical Sales & Services Supplier No. 497259994
RheBuild Kit for 9725, 9725I. Contains: Replacement Rotor Seals, Stator Face Assembly, Isolation Seal, Needle Guide, Needle Port Cleaner and Two Hex Nut Keys
Qty/Pkg: EACH