Learn More
Analytical Sales & Services RheBuild Kit for EV700-107 & EV750-107 (44750-1074 & 44700-1074)
Catalog No. 5000806916
Encompass
RheBuild Kit for EV700-107 and EV750-107 (44750-1074 and 44700-1074) Contains: Replacement Rotor Seals, Stator Face Assembly, Isolation Seal, Needle Guide, Needle Port Cleaner and Two Hex Nut Keys
Catalog No. 50-008-06916 Supplier Analytical Sales & Services Supplier No. 475079994
RheBuild Kit for EV700-107 and EV750-107 (44750-1074 and 44700-1074) Contains: Replacement Rotor Seals, Stator Face Assembly, Isolation Seal, Needle Guide, Needle Port Cleaner and Two Hex Nut Keys