Learn More
IDEX Health & Science Upchurch RHEBUILD KIT-MXT/HT/PD715-000
Catalog No. NC9064045
Encompass_Preferred
The RheBuild Kit maintenance kit reliably provides the normal wear parts, tools for disassembly/reassembly and instructions to maintain the precision performance your IDEX Health & Science valve.
Catalog No. NC9064045 Supplier IDEX Health & Science Upchurch Supplier No. 7150999
The RheBuild Kit maintenance kit reliably provides the normal wear parts, tools for disassembly/reassembly and instructions to maintain the precision performance your IDEX Health & Science valve.