Learn More
Analytical Sales & Services RheBuild Kit for RV7x3-100
Catalog No. 5000807025
Encompass
RheBuild Kit for RV7x3-100. Contains: Replacement Rotor Seals, Stator Face Assembly, Isolation Seal, Needle Guide, Needle Port Cleaner and Two Hex Nut Keys
Qty/Pkg: EACH
Catalog No. 50-008-07025 Supplier Analytical Sales & Services Supplier No. 475319994
RheBuild Kit for RV7x3-100. Contains: Replacement Rotor Seals, Stator Face Assembly, Isolation Seal, Needle Guide, Needle Port Cleaner and Two Hex Nut Keys
Qty/Pkg: EACH