Learn More
Cole-Parmer Idex RheBuild Kit for Valve Models 9010/9040; 1/EA
Supplier:  Cole-Parmer 0201577
Encompass
RheBulid kits contains normal wear parts, tools for disassembly/reassembly, and instructions to maintain your valves.
Catalog No. NC1681525