Learn More
Analytical Sales & Services RheFlex PEEK 1/16" Plugs
Catalog No. 5000805558
Encompass
RheFlex PEEK 1/16" Plugs.\n\nRheFlex tube fittings provide inert, non-metal connections for instrumentation. They can be used with all of our injectors, valves, and sample loops.
Catalog No. 50-008-05558 Supplier Analytical Sales & Services Supplier No. 460000904
RheFlex PEEK 1/16" Plugs.\n\nRheFlex tube fittings provide inert, non-metal connections for instrumentation. They can be used with all of our injectors, valves, and sample loops.