Learn More
Analytical Sales & Services RheFlex PEEK 1/16" Short Fittings Set
Catalog No. 5000805544
Encompass
RheFlex PEEK 1/16" Short Fittings Set, 10-32 RheFlex tube fittings provide inert, non-metal connections for instrumentation. They can be used with all of our injectors, valves, and sample loops.
Catalog No. 50-008-05544 Supplier Analytical Sales & Services Supplier No. 460000554
RheFlex PEEK 1/16" Short Fittings Set, 10-32 RheFlex tube fittings provide inert, non-metal connections for instrumentation. They can be used with all of our injectors, valves, and sample loops.