Learn More
Analytical Sales & Services RheFlex PEEK 1/8" Fittings Set
Catalog No. 5000807061
Encompass
RheFlex 1/8” PEEK Fittings Set\n\n• 5/16-24, 1/8" OD\n\nIncludes One Each Nut and Ferrule
Catalog No. 50-008-07061 Supplier Analytical Sales & Services Supplier No. 460000784
RheFlex 1/8” PEEK Fittings Set\n\n• 5/16-24, 1/8" OD\n\nIncludes One Each Nut and Ferrule