Learn More
Analytical Sales & Services RheFlex PEEK One-Piece Fitting for 1/32" OD Tubing, M4
Catalog No. 5000804980
Encompass
RheFlex One Piece PEEK Fitting, M4, for 1/32" OD Tubing
Catalog No. 50-008-04980 Supplier Analytical Sales & Services Supplier No. 460003374
RheFlex One Piece PEEK Fitting, M4, for 1/32" OD Tubing