Learn More
Analytical Sales & Services RheFlex PEEK One-Piece Fittings
Catalog No. 5000804727
Encompass
RheFlex One Piece Fingertight Fitting\n\n• 1-32 Threads\n\n• for 1/16" OD Tubing\n\n• PEEK\n\n• Color: Green\n\nQty/Pkg: 10
Catalog No. 50-008-04727 Supplier Analytical Sales & Services Supplier No. 460002824GN
Add to Cart
Add to Cart
RheFlex One Piece Fingertight Fitting\n\n• 1-32 Threads\n\n• for 1/16" OD Tubing\n\n• PEEK\n\n• Color: Green\n\nQty/Pkg: 10