Learn More
Analytical Sales & Services RheFlex PEEK One-Piece Fittings
Catalog No. 5000806954
Encompass
RheFlex One Piece Fingertight PEEK Fitting, 1-32 for 1/16" OD Tubing Color: Grey
Catalog No. 50-008-06954 Supplier Analytical Sales & Services Supplier No. 460002824GY
Add to Cart
Add to Cart
RheFlex One Piece Fingertight PEEK Fitting, 1-32 for 1/16" OD Tubing Color: Grey