Learn More
Analytical Sales & Services RheFlex PEEK One-Piece Fittings
Catalog No. 5000804317
Encompass
RheFlex One Piece Fingertight PEEK Fitting, 1-32 for 1/16" OD Tubing Color: Yellow
Catalog No. 50-008-04317 Supplier Analytical Sales & Services Supplier No. 460002824YL
RheFlex One Piece Fingertight PEEK Fitting, 1-32 for 1/16" OD Tubing Color: Yellow