Learn More
Selleck Chemical LLC Rheic Acid S2400-200mg
SDP
Catalog No. 503112720
Encompass_Preferred
Rhein (Monorhein NSC 38629 Rheinic) is an anthraquinone compound enriched in the fresh rhizome of Rheum coreanum Nakai showing anti-inflammation and antitumor activities Rhein is an FTO (mRNA N6-methyladenine demethylase) inhibitor and also inhibits AlkB repair enzymes
Catalog No. 50-311-2720 Supplier Selleck Chemical LLC Supplier No. S2400200MG
Add to Cart
Add to Cart
Rhein (Monorhein NSC 38629 Rheinic) is an anthraquinone compound enriched in the fresh rhizome of Rheum coreanum Nakai showing anti-inflammation and antitumor activities Rhein is an FTO (mRNA N6-methyladenine demethylase) inhibitor and also inhibits AlkB repair enzymes