Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Rhein

Catalog No. 27561150MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
50 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
27-561-150MG 50 mg
27-561-1250MG 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 27-561-150MG Supplier Sigma Aldrich Supplier No. 27561150MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Rhein

Specifications

CAS 478-43-3
Melting Point ≥300°C (lit.)
Molecular Formula C15H8O6
Linear Formula C15H8O6
Quantity 50 mg
MDL Number MFCD00009618
Synonym 4,5-Dihydroxyanthraquinone-2-carboxylic acid; 9,10-Dihydro-4,5-dihydroxy-9,10-dioxo-2-anthracenecarboxylic acid; Cassic acid; Chrysazin 3-carboxylic acid; Monorhein; Rheic acid; Rhubarb yellow
Molecular Weight (g/mol) 284.22
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.