Learn More
Cayman Chemical Rhein-8-glucoside, Purity: ≥98%, CAS: 34298-86-7, 10 mg
SDP
Catalog No. 502939082
Encompass_Preferred
An anthraquinone glycoside; increases the rate of sennoside A metabolism by mouse intestinal bacteria in vitro at 0.03-1 mM; increases the purgative activity of sennoside A in mice in a dose-dependent manner; inhibits PTP1B (IC50 = 11.5 μM); metabolized to rhein by intestinal bacteria
Catalog No. 50-293-9082 Supplier Cayman Chemical Supplier No. 2884410 MG
An anthraquinone glycoside; increases the rate of sennoside A metabolism by mouse intestinal bacteria in vitro at 0.03-1 mM; increases the purgative activity of sennoside A in mice in a dose-dependent manner; inhibits PTP1B (IC50 = 11.5 μM); metabolized to rhein by intestinal bacteria