Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Rhein 8-O-β-D-Glucopyranoside | 34298-86-7 | 99.8% | 446.36 | 5 MG
SDP

Catalog No. 5000393423
Encompass_Preferred

Rhein 8-O-β-D-Glucopyranoside is an anthraquinone glycoside found in rhubarb, known for its purgative activity.

  • Purity: 99.81%.
  • Appearance: Solid.
  • Color: Light yellow to yellow.
  • Initial source: Plants (Polygonaceae, Rheum palmatum L.).
  • Shipping: Room temperature in continental US; may vary elsewhere.
  • Storage: 4°C, protect from light.
  • Storage in solvent: -80°C for 6 months; -20°C for 1 month (protect from light).
  • Solubility (in vitro): 100 mg/mL (224.03 mM) in DMSO (requires ultrasonic, use newly opened DMSO due to hygroscopic nature).
  • Solubility (in vivo - protocol 1): ≥ 4.55 mg/mL (10.19 mM) in 10% DMSO, 40% PEG300, 5% Tween-80, 45% saline.
  • Solubility (in vivo - protocol 2): ≥ 2.5 mg/mL (5.60 mM) in 10% DMSO, 90% (20% SBE-β-CD in saline).

Catalog No. 50-003-93423 Supplier Medchemexpress LLC Supplier No. 5MGHYN6082
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Rhein 8-O-β-D-Glucopyranoside is an anthraquinone glycoside found in rhubarb, known for its purgative activity.

  • Purity: 99.81%.
  • Appearance: Solid.
  • Color: Light yellow to yellow.
  • Initial source: Plants (Polygonaceae, Rheum palmatum L.).
  • Shipping: Room temperature in continental US; may vary elsewhere.
  • Storage: 4°C, protect from light.
  • Storage in solvent: -80°C for 6 months; -20°C for 1 month (protect from light).
  • Solubility (in vitro): 100 mg/mL (224.03 mM) in DMSO (requires ultrasonic, use newly opened DMSO due to hygroscopic nature).
  • Solubility (in vivo - protocol 1): ≥ 4.55 mg/mL (10.19 mM) in 10% DMSO, 40% PEG300, 5% Tween-80, 45% saline.
  • Solubility (in vivo - protocol 2): ≥ 2.5 mg/mL (5.60 mM) in 10% DMSO, 90% (20% SBE-β-CD in saline).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.