Learn More
Strem, An Ascensus Company PentacarbonylchloroRe 98% 5g
SDP
Catalog No. 5090113073
Encompass
CAS# 14099-01-5. 5g. Rhenium pentacarbonyl chloride, 98%. MFCD00013296. Molecular Weight: 361.71. Molecular Formula: Re(CO)5Cl. Color/form: off-white xtl. Strem# 75-4000. http://www.strem.com/catalog/v/75-4000/
Catalog No. 50-901-13073 Supplier Strem, An Ascensus Company Supplier No. 7540005G
CAS# 14099-01-5. 5g. Rhenium pentacarbonyl chloride, 98%. MFCD00013296. Molecular Weight: 361.71. Molecular Formula: Re(CO)5Cl. Color/form: off-white xtl. Strem# 75-4000. http://www.strem.com/catalog/v/75-4000/