Learn More
Thermo Scientific™ Rhenium rod, 4.7-5mm (0.19-0.2in) dia, 99.97% (metals basis)
Specifications
Specifications
| CAS | 7440-15-5 |
| Physical Form | Solid |
| Quantity | 12.5 mm |
| Molecular Formula | Re |
| MDL Number | MFCD00011195 |
| InChI Key | WUAPFZMCVAUBPE-UHFFFAOYSA-N |
| SMILES | [Re] |
| IUPAC Name | rhenium |
| Molecular Weight (g/mol) | 186.21 |
| Percent Purity | 99.97% |
Safety and Handling
| ShelfLife | 30 Years |
| EINECSNumber | 231-124-5 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.