Learn More
Strem, An Ascensus Company CAS# 1314-28-9. Rhenium(VI) oxide (99.9%-Re). MFCD00016306. 1G
SDP
Catalog No. 5090113070
Encompass
CAS# 1314-28-9. 1G. Rhenium(VI) oxide (99.9%-Re). MFCD00016306. Molecular Formula: ReO3. Molecular Weight: 234.31. Color/Form: purple xtl. Strem#: 75-2500 www.strem.com/catalog/v3/75-2500/
Catalog No. 50-901-13070 Supplier Strem, An Ascensus Company Supplier No. 7525001G
CAS# 1314-28-9. 1G. Rhenium(VI) oxide (99.9%-Re). MFCD00016306. Molecular Formula: ReO3. Molecular Weight: 234.31. Color/Form: purple xtl. Strem#: 75-2500 www.strem.com/catalog/v3/75-2500/