Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Rhenium(Vii) Oxide

Catalog No. 2619555G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
26-195-55G 5 g
21-660-725ML 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 26-195-55G Supplier Sigma Aldrich Supplier No. 2619555G
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Rhenium(Vii) Oxide

Specifications

Boiling Point 360°C
Melting Point 220°C
CAS 1314-68-7
Quantity 5 g
Molecular Formula O7Re2
Linear Formula Re2O7
MDL Number MFCD00011199
Synonym Dirhenium heptoxide; Dirhenium heptoxide; Rhenium heptoxide
Molecular Weight (g/mol) 484.41 g/mol
Density 6.103 g/mL at 25°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.