Learn More
Strem, An Ascensus Company CAS# 1314-68-7. 1g. Rhenium(VII) oxide (99.99%-Re) PURATREM. MFCD00011199
SDP
Catalog No. 5090116331
Encompass
CAS# 1314-68-7. 1g. Rhenium(VII) oxide (99.99%-Re) PURATREM. MFCD00011199. Molecular Weight: 484.40. Molecular Formula: Re2O7. Color/form: yellow to pale green pwdr. Strem# 93-7505. http://www.strem.com/catalog/v/93-7505/
Catalog No. 50-901-16331 Supplier Strem, An Ascensus Company Supplier No. 9375051G
CAS# 1314-68-7. 1g. Rhenium(VII) oxide (99.99%-Re) PURATREM. MFCD00011199. Molecular Weight: 484.40. Molecular Formula: Re2O7. Color/form: yellow to pale green pwdr. Strem# 93-7505. http://www.strem.com/catalog/v/93-7505/