Specifications
| Material | Stainless Steel Valve |
| For Use With (Application) | Liquid Chromatography |
| Description | Fitting: 10-32 female Rheodyne valve ports; 5,000 psi max. pressure (350 bar) (factory set) |
Rheodyne is a registered trademark of IDEX Health & Science, LLC