Learn More
Waters Corp Waters Rheodyne 3725 Rebuild Kit, HPLC, 1 pk
Catalog No. 50791547 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This Performance Maintenance Kit is designed for the 3725 Rheodyne injector. It contains all the necessary parts to ensure the injector operates at peak performance.
  • Contains all necessary parts for peak performance
  • Designed for HPLC systems
  • Includes one rotor
  • Includes one rotor seal
  • Includes two allen wrenches
  • Comes as 1 unit per package
  • Replacement is as required
Catalog No. 50-791-547 Supplier Waters Corp Supplier No. 201000116
This Performance Maintenance Kit is designed for the 3725 Rheodyne injector. It contains all the necessary parts to ensure the injector operates at peak performance.
  • Contains all necessary parts for peak performance
  • Designed for HPLC systems
  • Includes one rotor
  • Includes one rotor seal
  • Includes two allen wrenches
  • Comes as 1 unit per package
  • Replacement is as required