Learn More
Waters Corp Waters Rheodyne 7725I Manual Injector, 1 to 20 μL, 6000 psi
Catalog No. 50793522 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
A manual injector from Waters, model Rheodyne 7725I, with 316 stainless steel and TFE wetted materials, capable of 1 to 20 μL injections and rated for 6000 psi pressure.
  • Wetted surface material includes 316 stainless steel and TFE.
  • Enables sample injections from 1 μL to 20 μL without a loop change.
  • Features a pressure limit of 6000 psi (420 kg/cm2).
  • Syringe needle measures 2.0 in. (5.08 cm) in length and 0.028 in. (0.071 cm) in outer diameter (22 gauge).
Catalog No. 50-793-522 Supplier Waters Corp Supplier No. WAT025248
A manual injector from Waters, model Rheodyne 7725I, with 316 stainless steel and TFE wetted materials, capable of 1 to 20 μL injections and rated for 6000 psi pressure.
  • Wetted surface material includes 316 stainless steel and TFE.
  • Enables sample injections from 1 μL to 20 μL without a loop change.
  • Features a pressure limit of 6000 psi (420 kg/cm2).
  • Syringe needle measures 2.0 in. (5.08 cm) in length and 0.028 in. (0.071 cm) in outer diameter (22 gauge).