Learn More
Waters Corp Rheodyne 8125 Rebuild kit
Supplier:  Waters Corp 201000120
Encompass_Preferred
The Waters Performance Maintenance Kit for the 8125 Rheodyne injector contains all the Waters Quality Parts necessary to keep your 8125 Rheodyne injector operating at peak performance. The Waters Performance Maintenance Kit for the 8125 injector includes the following parts: One rotor, one rotor seal, and two allen wrenches.
Catalog No. 50-791-551