Learn More
Waters Corp Waters Rebuild Kit, Prep Injection Valve, HPLC
Catalog No. 50792554 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This rebuild kit facilitates the replacement of the rotor seal and stator face assembly in an injection valve.
  • Involves changing the rotor seal and stator face assembly.
  • Designed for HPLC systems.
  • Replacement is recommended as required.
  • Contains 1 unit per package.
Catalog No. 50-792-554 Supplier Waters Corp Supplier No. 700001331
This rebuild kit facilitates the replacement of the rotor seal and stator face assembly in an injection valve.
  • Involves changing the rotor seal and stator face assembly.
  • Designed for HPLC systems.
  • Replacement is recommended as required.
  • Contains 1 unit per package.