Learn More
Waters Corp Waters Injection Valve, 7725i, 7-Port, Stainless Steel
Catalog No. 50793655 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The 7725i injection valve is a stainless steel, seven-port valve designed to support and maintain systems. It serves as an option for the 616 LC System and is also integrated into the Preparative Chromatography Rack for the 25X5 series pumps.
  • Spare parts kits
  • Model 7725i
  • Stainless steel seven-port injection valve
  • Sample is loaded through a built-in needle port in the front of the valve
  • Available as an option for the 616 LC System
  • Used in the Preparative Chromatography Rack for the 25X5 series pumps
Catalog No. 50-793-655 Supplier Waters Corp Supplier No. WAT055840
The 7725i injection valve is a stainless steel, seven-port valve designed to support and maintain systems. It serves as an option for the 616 LC System and is also integrated into the Preparative Chromatography Rack for the 25X5 series pumps.
  • Spare parts kits
  • Model 7725i
  • Stainless steel seven-port injection valve
  • Sample is loaded through a built-in needle port in the front of the valve
  • Available as an option for the 616 LC System
  • Used in the Preparative Chromatography Rack for the 25X5 series pumps