Learn More
MilliporeSigma™ Supelco™ Rheodyne™ Injector, Rheodyne™, 7725i
Catalog No. 111006491 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Max. Pressure:
7000 psi
Catalog No. Max. Pressure
11-100-6491 7000 psi
1 options
Catalog No. 11-100-6491 Supplier MilliporeSigma™ Supelco™ Supplier No. 57621
Add to Cart
Add to Cart
Internal position sensing switch, Manual, Number of ports: 6 and Number of positions: 2
Rheodyne™ Injector model is a stainless steel version and comprises of a six-port sample injection valves in which the sample is loaded through a built-in needle port in the front of the valve.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Liquid Chromatography
Max. Pressure 7000 psi
Description 7,000 psi max. pressure
Rheodyne is a registered trademark of IDEX Health & Science, LLC