Learn More
MilliporeSigma™ Supelco™ Rheodyne™ Model 3725i Preparative Sample Injector
Catalog No. 111002946 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Material:
PEEK
Catalog No. Material
11-100-2946 PEEK
1 options
Catalog No. 11-100-2946 Supplier MilliporeSigma™ Supelco™ Supplier No. 57461
Add to Cart
Add to Cart
Manual, Number of ports: 6 and Number of positions: 2
Model 3725i sample injectors for 1-10 cm I.D. HPLC columns combine the ease of use and versatility of Rheodyne′s analytical-scale injectors with the ability to handle large samples and high flow rates. Rugged and easily maintained, in a choice of stainless steel or PEEK flowpaths. Includes 10 mL sample loop, 1/8 in. fittings for all ports, 5 cm large-bore syringe needle (16 gauge/0.65 in. OD) for rapid injection of large samples, needle port cleaner, two vent tubes, wrenches, mounting screws, and instructions.
TRUSTED_SUSTAINABILITY
Specifications
Material PEEK
For Use With (Application) Liquid Chromatography
Rheodyne is a registered trademark of IDEX Health & Science, LLC