Learn More
MilliporeSigma™ Supelco™ Rheodyne™ Model 7000L Stream Switching Valve, Large Bore
Catalog No. 111003151 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Material:
Stainless Steel Valve
Catalog No. Material
11-100-3151 Stainless Steel Valve
1 options
Catalog No. 11-100-3151 Supplier MilliporeSigma™ Supelco™ Supplier No. 54479
Add to Cart
Add to Cart
Manual, Number of ports: 6 and Number of positions: 2
The 2-position 7000L Stream Switching Valve has a flow passage of 1 mm (0.040 in.).
TRUSTED_SUSTAINABILITY
Specifications
Material Stainless Steel Valve
For Use With (Application) Liquid Chromatography
Description Fitting: 10-32 Rheodyne thread; 5,000 psi max. pressure (350 bar) (factory set)
Rheodyne is a registered trademark of IDEX Health & Science, LLC