Learn More
MilliporeSigma™ Supelco™ Rheodyne™ Model 9725 Injector
Catalog No. 111003050 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
For Use With (Application):
Liquid Chromatography
Catalog No. For Use With (Application)
11-100-3050 Liquid Chromatography
1 options
Catalog No. 11-100-3050 Supplier MilliporeSigma™ Supelco™ Supplier No. 54432
Add to Cart
Add to Cart
Manual, Number of ports: 6, Number of positions: 2
The Model 9725 injector is ideal for both HPLC and soft gel LC purification procedures for biomolecules. Performance of this 6-port rotary injector is similar to that of the Model 7725 injector, but no metal contacts the samples. Model 9725 incorporates the patented make-before-break design (see Cat. No. 57620-U).
The 0.25 mm I.D. flow channels provide low dispersion when used with narrow I.D. columns, but prevent excessive flow resistance at flow rates used with larger analytical columns and preparative columns. Use with Rheodyne HPT fittings and high pressure PEEK tubing. Model 9725i includes a position-sensing switch.
Supplied with a 20 μL PEEK sample loop, Tefzel rotor seal, fittings for all ports, 22-gauge luer hub needle, needle port cleaner, two Tefzel vent tubes, wrenches, mounting screws and instructions.
Order additional sample loops separately.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Liquid Chromatography
Rheodyne is a registered trademark of IDEX Health & Science, LLC