Learn More
MilliporeSigma™ Supelco™ Rheodyne™ Model 9725i PEEK Injector
Catalog No. 111003041 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Material:
PEEK
Catalog No. Material
11-100-3041 PEEK
1 options
Catalog No. 11-100-3041 Supplier MilliporeSigma™ Supelco™ Supplier No. 54632
Add to Cart
Add to Cart
Manual with MBB and switch, Number of ports: 6, Number of positions: 2
The 9725i contains a switch and has the make before break (MBB). With the MBB, flow is not interrupted when switching from load to inject.
TRUSTED_SUSTAINABILITY
Specifications
Material PEEK
For Use With (Application) Liquid Chromatography
Description Fitting: 10-32 Rheodyne thread; 5,000 psi max. pressure
Rheodyne is a registered trademark of IDEX Health & Science, LLC