Learn More
Waters Corp Waters Corporation Rebuild Kit, Stainless Steel, 2-Position, 6-Port
Catalog No. 50792552 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This rebuild kit is designed for a stainless steel 2-position, 6-port motorized valve. It includes essential normal wear parts and tools required for disassembly and reassembly, ensuring proper maintenance of your system.
  • Rebuild kit type.
  • For HPLC systems.
  • Replacement as required.
  • One unit per package.
Catalog No. 50-792-552 Supplier Waters Corp Supplier No. 700001328
This rebuild kit is designed for a stainless steel 2-position, 6-port motorized valve. It includes essential normal wear parts and tools required for disassembly and reassembly, ensuring proper maintenance of your system.
  • Rebuild kit type.
  • For HPLC systems.
  • Replacement as required.
  • One unit per package.