Learn More
MilliporeSigma™ Supelco™ Rheodyne™ Rotor Seal for Model 7125/7725/7725i Injector
Catalog No. 111010426 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Material:
Tefzel™
Vespel
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
11-101-0426 Tefzel™
11-101-0354 Vespel
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-0426 Supplier MilliporeSigma™ Supelco™ Supplier No. 57633
Add to Cart
Add to Cart
Rotor Seal for Model 7125/7725/7725i Injector is an accessory for use with mainly Rheodyne™ model 7725 and 7725i injectors for operation at pH 0-14.
TRUSTED_SUSTAINABILITY
Specifications
Material Tefzel™
For Use With (Application) Liquid Chromatography
Rheodyne is a registered trademark of IDEX Health & Science, LLC