Learn More
Waters Corp Waters Rheodyne Prep Valve Assembly, 7-port, 3-position, Column/Fluidics Organizer
Catalog No. 50792638 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This prep valve assembly features a 7-port, 3-position configuration. It is designed to function as a column/fluidics organizer or flow straightener within high-performance liquid chromatography (HPLC) systems.
  • Functions as a 7-port, 3-position prep valve.
  • Designed as a column/fluidics organizer.
  • Suitable for use as a flow straightener.
Catalog No. 50-792-638 Supplier Waters Corp Supplier No. 700001830
This prep valve assembly features a 7-port, 3-position configuration. It is designed to function as a column/fluidics organizer or flow straightener within high-performance liquid chromatography (HPLC) systems.
  • Functions as a 7-port, 3-position prep valve.
  • Designed as a column/fluidics organizer.
  • Suitable for use as a flow straightener.